Merck
  • A General Light-Driven Organocatalytic Platform for the Activation of Inert Substrates.
A General Light-Driven Organocatalytic Platform for the Activation of Inert Substrates.
Angewandte Chemie (International ed. in English) (2023-06-16)
Shuo Wu, Florian Schiel, Paolo Melchiorre
ABSTRACT
Due to their strong covalent bonds and low reduction potentials, activating inert substrates is challenging. Recent advances in photoredox catalysis offered a number of solutions, each of which useful for activating specific inert bonds. Developing a general catalytic platform that can consistently target a broad range of inert substrates would be synthetically useful. Herein, we report a readily available indole thiolate organocatalyst that, upon excitation with 405 nm light, acquires a strongly reducing power. This excited-state reactivity served to activate, by single-electron reduction, strong C-F, C-Cl, and C-O bonds in both aromatic and aliphatic substrates. This catalytic platform was versatile enough to promote the reduction of generally recalcitrant electron-rich substrates (Ered <-3.0 V vs SCE), including arenes that afforded 1,4-cyclohexadienes. The protocol was also useful for the borylation and phosphorylation of inert substrates with a high functional group tolerance. Mechanistic studies identified an excited-state thiolate anion as responsible of the highly reducing reactivity.
Sigma-Aldrich
1-Methylindoline-2-thione, ≥95%