Merck
  • Enzymatic synthesis of (R)-cyanohydrins by a novel (R)-oxynitrilase from Vicia sativa L.
Enzymatic synthesis of (R)-cyanohydrins by a novel (R)-oxynitrilase from Vicia sativa L.
Biotechnology letters (2006-10-10)
Shi-Qing Han, Ping-Kai Ouyang, Ping Wei, Yi Hu
ABSTRACT
The defatted seed meal of common vetch (Vicia sativa L.) served as a source of (R)-oxynitrilase which catalyzed the enantioselective addition of HCN to aromatic, heteroaromatic, fluoro-substituted aromatic aldehydes to produce the corresponding enantiomeric pure cyanohydrins in yields of 52-100% and 88-99% ee at 12 degrees C under micro-aqueous medium (diisopropyl ether) without addition of buffer solution.