Merck
  • A new mutation in the regulatory domain of the myophosphorylase gene affecting protein dimer contact.
A new mutation in the regulatory domain of the myophosphorylase gene affecting protein dimer contact.
Muscle & nerve (1999-07-27)
J Gamez, R Fernandez, C Bruno, A L Andreu, C Cervera, C Navarro, S Schwartz, S Dimauro
ABSTRACT
We have identified a novel missense mutation in the myophosphorylase gene in a Spanish patient with McArdle's disease. The patient was homozygous for a T-to-C transition at codon 115 (L115P) in exon 3, which changed an encoded leucine (CUG) to a proline (CCG). This is the first mutation to be described in exon 3 and in a protein domain related to dimer contact. These data further emphasize the importance of private mutations in McArdle's disease, some of which are associated with specific ethnic groups.
Sigma-Aldrich
Phosphorylase b from rabbit muscle, For use as a marker in SDS-PAGE
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization
Sigma-Aldrich
Phosphorylase a from rabbit muscle, lyophilized powder, 20-30 units/mg protein