Merck
  • Regulation of phosphoglucomutase 1 phosphorylation and activity by a signaling kinase.
Regulation of phosphoglucomutase 1 phosphorylation and activity by a signaling kinase.
Oncogene (2004-09-21)
Anupama Gururaj, Christopher J Barnes, Ratna K Vadlamudi, Rakesh Kumar
ABSTRACT
We have identified a novel mechanism of cross-talk between cell signaling and metabolic pathways, whereby the signaling kinase p21-activated kinase 1 (Pak1) binds to, phosphorylates and enhances the enzymatic activity of phosphoglucomutase 1 (PGM), an important regulatory enzyme in cellular glucose utilization and energy homeostasis. Pak1 and PGM were colocalized in model cell systems and showed functional interactions in a physiological setting. Strong direct interaction of PGM with Pak1 but not Pak2, Pak3, or Pak4 was observed. PGM binding was within 75-149 amino acids (aa) of Pak1, while Pak1 binding to PGM was in the N-terminal 96 aa. Pak1-mediated phosphorylation of PGM selectively on threonine 466 significantly increased PGM enzymatic activity and could be blocked by transfection with a dominant-negative Pak1 expression vector and by Pak1-specific small inhibitory RNA. Stable transfection of PGM into PGM-deficient K562 leukemia cells further demonstrated the role of Pak1 in regulating PGM activity. The results presented here provide new evidence that the cell signaling kinase Pak1 is a novel regulator of glucose metabolism through its phosphorylation and regulation of PGM activity. These findings suggest a new mechanism whereby growth factor signaling may coordinately integrate metabolic regulation with established signaling functions of cell cycle regulation and cell growth.
Sigma-Aldrich
Phosphoglucomutase from rabbit muscle, ammonium sulfate suspension, ≥100 units/mg protein