Merck
  • Synthesis and structural characterization of binuclear half-sandwich iridium, rhodium and ruthenium complexes containing 4,4'-dipyridyldisulfide (4DPDS) ligands.
Synthesis and structural characterization of binuclear half-sandwich iridium, rhodium and ruthenium complexes containing 4,4'-dipyridyldisulfide (4DPDS) ligands.
Dalton transactions (Cambridge, England : 2003) (2010-07-02)
Ying-Feng Han, Hao Li, Yue Fei, Yue-Jian Lin, Wan-Zheng Zhang, Guo-Xin Jin
ABSTRACT
Reactions of half-sandwich iridium and rhodium complexes [Cp*MCl2]2 (1a: M = Ir, 1b: M = Rh) and the half-sandwich ruthenium complex [(p-cymene)RuCl2]2 (1c) with 4DPDS gave the corresponding binuclear complexes [{Cp*MCl2]2}(m-4DPDS) (2a: M = Ir, 2b: M = Rh) and [{(p-cymene)RuCl2]2}(m-4DPDS) (2c), which can be converted into binuclear metallamacrocycles. The metallamacrocycles [Cp*M(m-4DPDS)Cl]2(2+) (3a: M = Ir, 3b: M = Rh) and [(p-cymene)Ru(m-4DPDS)Cl]2(2+) (3c) are composed of two half-sandwich units and two 4DPDS ligands with the same chirality. The macrocycle 3c is chiral and changes its shape slightly depending on the guests accommodated above and below the cavities.
Sigma-Aldrich
Aldrithiol-4, 98%