Merck
  • Giant basal cell carcinoma of the breast mimicking paget's disease: complete remission after photodynamic therapy.
Giant basal cell carcinoma of the breast mimicking paget's disease: complete remission after photodynamic therapy.
Onkologie (2010-10-27)
Nadia C Mikhaimer, Katharina C Kähler, Thomas Schwarz, Christoph Mundhenke, Axel Hauschild
ABSTRACT
Basal cell carcinoma (BCC) in the nipple area is extremely rare and can be mistaken for Paget's disease. A 47-year-old woman complained of a slowly growing, scaly erythematous plaque around her right nipple which had developed over several years. Paget's disease of the breast was suspected. Biopsies revealed a superficial BCC. The maximum tumor thickness was measured as 1 mm. Considering the superficial character of the tumor, photodynamic therapy (PDT) as a non-invasive approach was performed. Complete remission (CR) with an excellent cosmetic result was achieved after 4 PDT sessions. In the follow-up, CR with no signs of a local recurrence was evident after more than 24 months. In similar superficial cases of BCCs, non-invasive procedures such as PDT can be discussed as an alternative option to radical surgery or radiotherapy, with an excellent aesthetic outcome in carefully selected cases.
Sigma-Aldrich
5-Aminovaleric acid, 97%