Merck
Short-term regulation of adiponectin secretion in rat adipocytes.
Physiological research (2011-03-16)
T Szkudelski, L Nogowski, K Szkudelska
ABSTRACT
Adiponectin belongs to the group of biologically active substances secreted by adipocytes and referred to as adipokines. Disturbances in its secretion and/or action are thought to be involved in the pathogenesis of some metabolic diseases. However, regulation of adiponectin secretion is poorly elucidated. In the present study, short-term regulation of adiponectin secretion in primary rat adipocytes was investigated. Isolated rat adipocytes were incubated in Krebs-Ringer buffer containing 5 mM glucose and insulin alone or in the combination with epinephrine, dibutyryl-cAMP, adenosine A(1) receptor antagonist (DPCPX), palmitate, 2-bromopalmitate or inhibitor of mitochondrial electron transport (rotenone). Adipocyte exposure for 2 h to insulin (1-100 nM) significantly increased secretion of adiponectin compared with secretion observed without insulin. Furthermore, secretion of adiponectin from adipocytes incubated with glucose and insulin was reduced by 1 and 2 microM epinephrine, but not by 0.25 and 0.5 microM epinephrine. Under similar conditions, 1 and 2 mM dibutyryl-cAMP substantially diminished secretion of adiponectin, whereas 0.5 mM dibutyryl-cAMP was ineffective. Secretion of adiponectin was found to be effectively decreased by DPCPX. Moreover, adipocyte exposure to rotenone also resulted in a substantial diminution of secretory response of adipocytes incubated for 2 h with glucose and insulin. It was also demonstrated that palmitate and 2-bromopalmitate (0.06-0.5 mM) failed to affect secretion of leptin. The obtained results indicated that in short-term regulation of adiponectin secretion, insulin and epinephrine exert the opposite effects. These effects appeared as early as after 2 h of exposure. Moreover, deprivation of energy or blockade of adenosine action substantially decreased secretion of adiponectin.
Sigma-Aldrich
2-Bromohexadecanoic acid, ~97%
Sigma-Aldrich
2-Bromohexadecanoic acid, ≥99.0% (GC)