Merck
  • Efficiency enhancement of organic solar cells by fabricating periodic surface textures using direct laser interference patterning.
Efficiency enhancement of organic solar cells by fabricating periodic surface textures using direct laser interference patterning.
Advanced materials (Deerfield Beach, Fla.) (2012-03-10)
Lars Müller-Meskamp, Yong Hyun Kim, Teja Roch, Simone Hofmann, Reinhard Scholz, Sebastian Eckardt, Karl Leo, Andrés Fabián Lasagni
ABSTRACT
Direct laser interference patterning (DLIP) is used to fabricate large area, two-dimensional periodic surface patterns on polyethylene terephthalate (PET) substrates to enhance the performance of ZnPc:C60 solar cells by light concentration in the absorber layer. Comparing the power conversion efficiencies to the reference cell on flat PET, a relative increase of 21% is observed for the hexagonal pattern with 0.7 μm period, depicted in the figure.
Sigma-Aldrich
Zinc phthalocyanine, Dye content 97 %