Merck
  • Chronic clomipramine treatment restores hippocampal expression of glial cell line-derived neurotrophic factor in a rat model of depression.
Chronic clomipramine treatment restores hippocampal expression of glial cell line-derived neurotrophic factor in a rat model of depression.
Journal of affective disorders (2012-06-05)
Qiong Liu, Hai-Yan Zhu, Bing Li, Yan-Qing Wang, Jin Yu, Gen-Cheng Wu
ABSTRACT
Because there is evidence that certain neurotrophic factors are involved in depression and the mechanism of antidepressant treatment, it is hypothesized that neurotrophic factors may also play a functional role in the etiology of depression and treatment. Glial cell line-derived neurotrophic factor (GDNF) is a member of the transforming growth factor (TGF-β)-super-family. We performed a study to assess the impact of chronic unpredictable stress (CUS) and clomipramine treatment on GDNF expression in the rat hippocampus. Using a rat model of CUS-induced depression, we administered clomipramine, one of the typical antidepressants, every day for 3 weeks starting 2 weeks after the beginning of the experiment. GDNF level in the hippocampus was detected by immunohistochemsitry, Western blot analysis, and reverse transcription-polymerase chain reaction (RT-PCR). Behavioral changes were measured by forced swimming test (FST) and open field test (OFT). Animals exposed to CUS showed depression-like behavior and exhibited a significant decrease in GDNF expression in the hippocampus. Chronic clomipramine treatment reversed the behavioral deficits and the decrease in GDNF levels induced by CUS. The relatively small number of the depression-model rats may cause some bias of behavioral tests. In our study, chronic clomipramine treatment restored GDNF expression in the hippocampus of CUS-induced depression rats, suggesting that GDNF is involved in the behavioral responses to antidepressants. The beneficial effects of clomipramine suggest that GDNF may be a viable target for new antidepressant drug development.
Sigma-Aldrich
Clomipramine hydrochloride, ≥98% (HPLC), powder