Merck
  • Discovery of novel 2-(3-(2-chlorophenyl)pyrazin-2-ylthio)-N-arylacetamides as potent HIV-1 inhibitors using a structure-based bioisosterism approach.
Discovery of novel 2-(3-(2-chlorophenyl)pyrazin-2-ylthio)-N-arylacetamides as potent HIV-1 inhibitors using a structure-based bioisosterism approach.
Bioorganic & medicinal chemistry (2012-10-27)
Peng Zhan, Wenmin Chen, Zhenyu Li, Xiao Li, Xuwang Chen, Ye Tian, Christophe Pannecouque, Erik De Clercq, Xinyong Liu
ABSTRACT
The present work is an extension of our ongoing efforts towards the development and identification of new molecules with anti-HIV activity which have previously led to the discovery of arylazolylthioacetanilides as highly active NNRTIs. In this article, a series of 2-2-(3-(2-chlorophenyl)pyrazin-2-ylthio)-N-arylacetamide derivatives were synthesized and evaluated for in vitro anti-HIV activity. Most of the tested compounds exhibited moderate activities against wild-type HIV-1. Among them, compound 6k showed significant activity against wild-type HIV-1 with an EC(50) value of 1.7μM, along with moderate activity against wild-type reverse transcriptase (RT). The preliminary structure-activity relationship (SAR) and docking calculations of this new series of compounds were also investigated, which may help designing more potent molecules.
Sigma-Aldrich
Thioacetamide, reagent grade, 98%
Sigma-Aldrich
Thioacetamide, ACS reagent, ≥99.0%