Merck
  • Effect of cadmium chloride on secretion of 17β-estradiol by the ovarian follicles of common carp, Cyprinus carpio.
Effect of cadmium chloride on secretion of 17β-estradiol by the ovarian follicles of common carp, Cyprinus carpio.
General and comparative endocrinology (2012-11-14)
Sumana Das, Dilip Mukherjee
ABSTRACT
Cadmium (Cd(2+)) is a common environmental pollutant present in wastes associated with mining, smelting and electroplating. It is a major constituent of the tobacco smoke. Exposure of this heavy metal has been linked to wide range of detrimental effects on mammalian reproduction particularly on ovarian steroidogenesis. Low doses of Cd(2+) are reported to stimulate ovarian luteal progesterone synthesis whereas high doses inhibited it. Cd(2+) exposure is also reported to inhibit gonadal function in fish. In the present study the effects of cadmium chloride (CdCl(2)) on the secretion of gonadotropin-induced 17β-estradiol was examined in female common carp Cyprinus carpio. Vitellogenic stage fish were exposed to physiological safe dose of CdCl(2) for 0, 24, 48 and 96 h and serum and ovarian 17β-estradiol levels were estimated. In the in vitro experiments, vitellogenic follicles were incubated with CdCl(2) and a dose- and time-dependent effects on steroid production were estimated induced by LH. Exposure of fish with CdCl(2) gradually attenuated serum and ovarian 17β-estradiol levels with increasing time and maximum inhibition was noticed after 96 h. Administration of CdCl(2) to the incubations significantly inhibited LH-induced release of 17β-estradiol in vitro. To clarify the mechanism of attenuated production of 17β-estradiol, in vitro effects of CdCl(2) on LH induced P450 aromatase activity (conversion of testosterone to 17β-estradiol) and cytochrome P450arom gene expression in carp ovarian follicles were evaluated. Results show that LH-stimulated P450 aromatase activity and P450arom gene expression in ovarian follicles were significantly inhibited by CdCl(2). The present study further demonstrated that LH-induced stimulation of ovarian steroidogenic factor-1 (SF-1) which activates aromatase enzyme, is strongly inhibited by cadmium chloride treatment.
Sigma-Aldrich
Cadmium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cadmium chloride