Merck
  • Improved synthesis of the peripheral benzodiazepine receptor ligand [11C]DPA-713 using [11C]methyl triflate.
Improved synthesis of the peripheral benzodiazepine receptor ligand [11C]DPA-713 using [11C]methyl triflate.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2006-01-24)
C Thominiaux, F Dollé, M L James, Y Bramoullé, H Boutin, L Besret, M-C Grégoire, H Valette, M Bottlaender, B Tavitian, Ph Hantraye, S Selleri, M Kassiou
ABSTRACT
Recently, the pyrazolopyrimidine, [11C] N,N-Diethyl-2-[2-(4-methoxyphenyl)-5,7-dimethylpyrazolo[1,5-a]pyrimidin-3-yl]acetamide (DPA-713) has been reported as a new promising marker for the study of peripheral benzodiazepine receptors with positron emission tomography. In the present study, DPA-713 has been labelled from the corresponding nor-analogue using [11C]methyl triflate (CH3OTf). Conditions for HPLC were also modified to include physiological saline (aq. 0.9% NaCl)/ethanol:60/40 as mobile phase making it suitable for injection. The total time of radiosynthesis, including HPLC purification, was 18-20 min. This reported synthesis of [11C]DPA-713, using [11C]CH3OTf, resulted in an improved radiochemical yield (30-38%) compared to [11C]methyl iodide (CH3I) (9) with a simpler purification method. This ultimately enhances the potential of [11C]DPA-713 for further pharmacological and clinical evaluation. These improvements make this radioligand more suitable for automated synthesis which is of benefit where multi-dose preparations and repeated syntheses of radioligand are required.
Sigma-Aldrich
Methyl trifluoromethanesulfonate, ≥98%