Merck
  • Two novel mutations in splice donor sites of CYP11B1 in congenital adrenal hyperplasia due to 11beta-hydroxylase deficiency.
Two novel mutations in splice donor sites of CYP11B1 in congenital adrenal hyperplasia due to 11beta-hydroxylase deficiency.
Endocrine research (2001-02-24)
O Chabre, S Portrat-Doyen, J Vivier, Y Morel, G Defaye
ABSTRACT
We present an in vivo and in vitro study of congenital adrenal hyperplasia in a patient with 11beta-hydroxylase deficiency. Genetic analysis showed two new base substitutions of CYP11B1, a conservative transition at the last base of exon 5, and a IVS8+4A-->G transition in intron 8. Difficulties with suppressive therapy resulted in severe hypertension. A laparoscopic adrenalectomy was decided which lead to normalization of blood pressure. In vitro, steroidogenesis by adrenal cells showed no measurable 11beta-hydroxylase activity. Analysis of CYP11B1 mRNA by RT-PCR and sequencing showed expression of a mRNA which lacked exon 8, presumably resulting from the intron 8 mutation. In addition a highly truncated mRNA was detected corresponding to exons 1, 2, 8, 9, with the loss of exons 3-7, presumably related to the exon 5 mutation. Western blot analysis showed a shorter CYP11B immunoreactive band of 43 kDa, consistent with truncation of exon 8. Thus adrenalectomy in this patient allowed effective treatment of severe hypertension and helped to understand the mechanisms of two novel mutations responsible for aberrant splicing of CYP11B1.
Sigma-Aldrich
21-Deoxycortisol