Merck
DBD dyes as fluorescent probes for sensing lipophilic environments.
Bioorganic & medicinal chemistry letters (2012-08-04)
Robert Wawrzinek, Pablo Wessig, Kristian Möllnitz, Jörg Nikolaus, Roland Schwarzer, Peter Müller, Andreas Herrmann
ABSTRACT
Small fluorescent organic molecules based on [1,3]dioxolo[4,5-f][1,3]benzodioxole (DBD) could be used as probes for lipophillic microenvironments in aqueous solutions by indicating the critical micelles concentration of detergents and staining cell organelles. Their fluorescence lifetime decreases drastically by the amount of water in their direct environment. Therefore they are potential probes for fluorescence lifetime imaging microscopy (FLIM).
Sigma-Aldrich
1,3-Benzodioxole, 99%