Merck
  • Further studies on the substrate spectrum of phytanoyl-CoA hydroxylase: implications for Refsum disease?
Further studies on the substrate spectrum of phytanoyl-CoA hydroxylase: implications for Refsum disease?
Journal of lipid research (2003-08-19)
Veerle Foulon, Stanny Asselberghs, Wendy Geens, Guy P Mannaerts, Minne Casteels, Paul P Van Veldhoven
ABSTRACT
Refsum disease is a peroxisomal disorder characterized by adult-onset retinitis pigmentosa, anosmia, sensory neuropathy, ataxia, and an accumulation of phytanic acid in plasma and tissues. Approximately 45% of cases are caused by mutations in phytanoyl-CoA hydroxylase (PAHX), the enzyme catalyzing the second step in the peroxisomal alpha-oxidation of 3-methyl-branched fatty acids. To study the substrate specificity of human PAHX, different 3-alkyl-branched substrates were synthesized and incubated with a recombinant polyhistidine-tagged protein. The enzyme showed activity not only toward racemic phytanoyl-CoA and the isomers of 3-methylhexadecanoyl-CoA, but also toward a variety of other mono-branched 3-methylacyl-CoA esters with a chain length down to seven carbon atoms. Furthermore, PAHX hydroxylated a 3-ethylacyl-CoA quite well, whereas a 3-propylacyl-CoA was a poor substrate. Hydroxylation of neither 2- or 4-methyl-branched acyl-CoA esters, nor long or very long straight-chain acyl-CoA esters could be detected. The results presented in this paper show that the substrate specificity of PAHX, with regard to the length of both the acyl-chain and the branch at position 3, is broader than expected. Hence, Refsum disease might be characterized by an accumulation of not only phytanic acid but also other 3-alkyl-branched fatty acids.
Sigma-Aldrich
2-Decanol, 98%