- Surface intermediate species of the 4-isobutylacetophenone adsorption-reaction over fosfotungstic Wells-Dawson heteropoly acid.
Surface intermediate species of the 4-isobutylacetophenone adsorption-reaction over fosfotungstic Wells-Dawson heteropoly acid.
Latin American Applied Research (2009)
Matkovic SR, et al.