Merck
Total synthesis of ambruticin.
Angewandte Chemie (International ed. in English) (2002-12-20)
Eun Lee, Seung Jib Choi, Hahn Kim, Hee Oon Han, Young Keun Kim, Sun Joon Min, Sung Hee Son, Sang Min Lim, Won Suk Jang
Sigma-Aldrich
L-(+)-Arabinose, 98%