Merck
Development of a new colorimetric assay for lipoxygenase activity.
Analytical biochemistry (2013-07-03)
Weiqiang Lu, Xue Zhao, Zhongyu Xu, Ningning Dong, Shien Zou, Xu Shen, Jin Huang
ABSTRACT
Lipoxygenases (LOXs) are a family of non-heme iron-containing dioxygenases that catalyze the hydroperoxidation of lipids, containing a cis,cis-1,4-pentadiene structure. A rapid and reliable colorimetric assay for determination of the activity of three human functional lipoxygenase isoforms (5-lipoxygenase, platelet 12-lipoxygenase, and 15-lipoxygenase-1) is developed in this article. In the new assay, LOX-derived lipid hydroperoxides oxidize the ferrous ion (Fe²⁺) to the ferric ion (Fe³⁺), the latter of which binds with thiocyanate (SCN⁻) to generate a red ferrithiocyanate (FTC) complex. The absorbance of the FTC complex can be easily measured at 480 nm. Because 5-LOX can be stimulated by many cofactors, the effects of its cofactors (Ca²⁺, ATP, dithiothreitol, glutathione, L-α-phosphatidylcholine, and ethylenediaminetetraacetic acid) on the color development of the FTC complex are also determined. The assay is adaptive for purified LOXs and cell lysates containing active LOXs. We use the new colorimetric assay in a 96-well format to evaluate several well-known LOX inhibitors, the IC₅₀ values of which are in good agreement with previously reported data. The reliability and reproducibility of the assay make it useful for in vitro screening for inhibitors of LOXs and, therefore, should accelerate drug discovery for clinical application.
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type I-B, lyophilized powder, ≥50,000 units/mg solid
Sigma-Aldrich
Lipoxidase from Glycine max (soybean), Type V, ammonium sulfate suspension, 500,000-1,000,000 units/mg protein