Merck
  • Fluorometric high-performance liquid chromatography of N-acetyl- and N-glycolylneuraminic acids and its application to their microdetermination in human and animal sera, glycoproteins, and glycolipids.
Fluorometric high-performance liquid chromatography of N-acetyl- and N-glycolylneuraminic acids and its application to their microdetermination in human and animal sera, glycoproteins, and glycolipids.
Analytical biochemistry (1987-07-01)
S Hara, Y Takemori, M Yamaguchi, M Nakamura, Y Ohkura
ABSTRACT
A simple, rapid, and highly sensitive high-performance liquid chromatographic method is described for the determination of N-acetyl- and N-glycolylneuraminic acids in human and animal sera, glycoproteins, and glycolipids. The neuraminic acids, released by acid hydrolysis of these biological samples, are converted in dilute sulfuric acid with 1,2-diamino-4,5-methylene-dioxybenzene, a fluorogenic reagent for alpha-keto acids, to highly fluorescent derivatives. The derivatives are separated within 12 min on a reversed-phase column (Radial-Pak cartridge C18) with an isocratic elution and detected fluorometrically. The detection limits are 25 fmol (7.7 pg) for N-acetylneuraminic acid and 23 fmol (7.5 pg) for N-glycolylneuraminic acid in a 10-microliter injection volume at a signal-to-noise ratio of 2. This method permits precise determination of the neuraminic acids in 5 microliter of human and animal sera or in 0.25-2.5 micrograms of glycoproteins and glycolipids.
Sigma-Aldrich
4,5-Methylenedioxy-1,2-phenylenediamine dihydrochloride, BioReagent, suitable for fluorescence, ≥97.0% (HPLC)