Merck
Pimozide: use in dermatology.
Dermatology online journal (2003-03-18)
Willem A van Vloten
ABSTRACT
Pimozide is widely used in psychiatry for chronic psychoses, schizophrenia, the syndrome of Gilles de la Tourette and to a certain extent, also in dermatology. The only dermatological indication is for delusions of parasitosis. Though there is a good rationale for using pimozide in this disease, the majority of the studies on pimozide in dermatology are uncontrolled trials and case reports.
Product Description
Pimozide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pimozide