Merck
Triclosan.
Dermatitis : contact, atopic, occupational, drug (2006-12-08)
Lauren Campbell, Matthew J Zirwas
ABSTRACT
Triclosan is a common antibacterial chemical currently in widespread use in household and health care-related products. Triclosan was first introduced in 1965 and has been marketed as cloxifenol, Irgasan CH 3565, and Irgasan DP 300. Its most common use is in antimicrobial hand soaps, but in the United States it can also be found in consumer products such as liquid dishwashing soaps, deodorants, and toothpastes. The concentrations used in products in the United States typically range from 0.15 to 0.3%. While allergy to triclosan-based products is uncommon, several cases of contact dermatitis secondary to triclosan have been reported.
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material