Merck
  • Enantioselective construction of functionalized thiopyrano-indole annulated heterocycles via a formal thio [3 + 3]-cyclization.
Enantioselective construction of functionalized thiopyrano-indole annulated heterocycles via a formal thio [3 + 3]-cyclization.
Organic letters (2014-12-18)
Xiang Chen, Zheng-Hang Qi, Shao-Yun Zhang, Ling-Pei Kong, Yong Wang, Xing-Wang Wang
ABSTRACT
A formal thio [3 + 3]-cyclization catalyzed by a DPEN-derived chiral thiourea has been reported for the construction of optically active thiopyrano-indole annulated heterocyclic compounds in high yields with excellent enantioselectivities. The high reactivity between indoline-2-thione (keto-S) and 2-benzylidenemalononitrile has also been supported by density functional theory (DFT) calculations.
Sigma-Aldrich
Thiourea, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiourea, ReagentPlus®, ≥99.0%
Sigma-Aldrich
1-Methylindoline-2-thione, ≥95%