Merck
  • Reduced expression of PTPRD correlates with poor prognosis in gastric adenocarcinoma.
Reduced expression of PTPRD correlates with poor prognosis in gastric adenocarcinoma.
PloS one (2014-11-21)
Dandan Wang, Leilei Wang, Jun Zhou, Jihong Pan, Wei Qian, Jiafang Fu, Genglin Zhang, Youming Zhu, Chunshan Liu, Chunliang Wang, Zongkun Jin, Ziqing He, Jianmei Wu, Bin Shi
ABSTRACT
PTPRD, encoding protein tyrosine phosphatases receptor type D, is located at chromosome 9p23-24.1, a loci frequently lost in many types of tumors. Recently, PTPRD has been proposed to function as a tumor suppressor gene. The current study aimed to investigate PTPRD expression and its prognostic significance in primary gastric adenocarcinoma. Quantitative real time reverse transcription PCR (qRT-PCR) and western blotting were used to examine PTPRD expression in paired gastric tumourous and paracancerous tissues. Compared with the matched normal gastric mucosa tissues, both the mRNA (P = 0.0138) and protein (P = 0.0093) expression of PTPRD in fresh surgical specimens were significantly reduced. Clinicopathological and prognostic roles of PTPRD in gastric adenocarcinoma were investigated using immunohistochemistry with 513 paraffin-embedded gastric adenocarcinoma tissue blocks. Statistical analysis revealed that reduced PTPRD expression was significantly associated with T stage (P = 0.004), TNM stage (P<0.001) and tumor size (P = 0.003). Furthermore, Kaplan-Meier survival analysis revealed that low expression of PTPRD significantly correlated with poor survival of gastric cancer patients (P<0.001). Cox regression analysis confirmed PTPRD expression as independent predictor of the overall survival of gastric cancer patients. The MTT assay determined the effects of PTPRD on cell proliferation of MGC803 and GES1 cell lines. Restoring PTPRD expression in MGC803 cells significantly inhibited their growth rate. Silencing PTPRD expression by siRNA treatment in GES1 significantly enhanced cell proliferation compared with mock siRNA treatment. Methylation analysis of PTPRD promoter CpG island in 3 primary GC samples showed one case with partial methylation. These results indicated that PTPRD is a candidate tumour suppressor in gastric cancer. Thus, PTPRD may play an important role in gastric tumorigenesis and serve as a valuable prognostic marker of gastric adenocarcinoma.
Sigma-Aldrich
MISSION® esiRNA, targeting human PTPRD