Merck
  • Epithelial protein lost in neoplasm modulates platelet-derived growth factor-mediated adhesion and motility of mesangial cells.
Epithelial protein lost in neoplasm modulates platelet-derived growth factor-mediated adhesion and motility of mesangial cells.
Kidney international (2014-04-04)
Haruko Tsurumi, Yutaka Harita, Hidetake Kurihara, Hidetaka Kosako, Kenji Hayashi, Atsuko Matsunaga, Yuko Kajiho, Shoichiro Kanda, Kenichiro Miura, Takashi Sekine, Akira Oka, Kiyonobu Ishizuka, Shigeru Horita, Motoshi Hattori, Seisuke Hattori, Takashi Igarashi
ABSTRACT
Mesangial cell migration, regulated by several growth factors, is crucial after glomerulopathy and during glomerular development. Directional migration requires the establishment of a polarized cytoskeletal arrangement, a process regulated by coordinated actin dynamics and focal adhesion turnover at the peripheral ruffles in migrating cells. Here we found high expression of the actin cross-linking protein EPLIN (epithelial protein lost in neoplasm) in mesangial cells. EPLIN was localized in mesangial angles, which consist of actin-containing microfilaments extending underneath the capillary endothelium, where they attach to the glomerular basement membrane. In cultured mesangial cells, EPLIN was localized in peripheral actin bundles at focal adhesions and formed a protein complex with paxillin. The MEK-ERK (extracellular signal-regulated kinase) cascade regulated EPLIN-paxillin interaction and induced translocalization of EPLIN from focal adhesion sites to peripheral ruffles. Knockdown of EPLIN in mesangial cells enhanced platelet-derived growth factor-induced focal adhesion disassembly and cell migration. Furthermore, EPLIN expression was decreased in mesangial proliferative nephritis in rodents and humans in vivo. These results shed light on the coordinated actin remodeling in mesangial cells during restorative remodeling. Thus, changes in expression and localization of cytoskeletal regulators underlie phenotypic changes in mesangial cells in glomerulonephritis.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Lima1
Sigma-Aldrich
MISSION® esiRNA, targeting human LIMA1