Merck
  • Cyanosilylation of benzaldehyde with TMSCN over perovskite-type oxide catalyst prepared by thermal decomposition of heteronuclear cyano complex precursors.
Cyanosilylation of benzaldehyde with TMSCN over perovskite-type oxide catalyst prepared by thermal decomposition of heteronuclear cyano complex precursors.
Research on Chemical Intermediates (2015)
Yamaguchi S, et al.