Merck
  • Silencing of TGIF1 in bone mesenchymal stem cells applied to the post-operative rotator cuff improves both functional and histologic outcomes.
Silencing of TGIF1 in bone mesenchymal stem cells applied to the post-operative rotator cuff improves both functional and histologic outcomes.
Journal of molecular histology (2015-03-19)
Jie Li, Liyang Chen, Lin Sun, Hua Chen, Yeqing Sun, Chaoyin Jiang, Biao Cheng
ABSTRACT
Stem cells have long been hypothesized to improve outcomes following rotator cuff repair. However, these cells must be signaled in order to do so. TGIF1 is a transcription factor that has been found to be down-regulated in cells involved in chondrogenesis. We therefore wished to examine whether stem cells expressing lower levels of TGIF1 could better improve outcomes following rotator cuff repair than stem cells expressing normal levels of TGIF1. Bone mesenchymal stem cells (BMSCs) were transduced with TGIF1 siRNA to suppress native TGIF1. Nontransduced BMSCs were also obtained for the control group. Following suprapinatus tendon repair, rats were either treated with transduced BMSCs or nontransduced BMSCs. Histologic and functional testing were performed on both groups. Rats treated with transduced TGIF1 siRNA BMSCs following suprapinatus repair expressed significantly higher levels of chondrogenic proteins at 4 weeks than rats treated with nontransduced BMSCs. Further, rats treated with BMSCs transduced with TGIF1 siRNA had both a significantly greater maximum load at failure and stiffness. Rats treated with transduced TGIF1 siRNA BMSCs following supraspinatus repair perform better both histologically and functionally at 4 weeks.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tgif1
Sigma-Aldrich
DL-Proline, ReagentPlus®, 99%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%