Merck
  • ZFP36L1 promotes monocyte/macrophage differentiation by repressing CDK6.
ZFP36L1 promotes monocyte/macrophage differentiation by repressing CDK6.
Scientific reports (2015-11-07)
Ming-Tai Chen, Lei Dong, Xin-Hua Zhang, Xiao-Lin Yin, Hong-Mei Ning, Chao Shen, Rui Su, Feng Li, Li Song, Yan-Ni Ma, Fang Wang, Hua-Lu Zhao, Jia Yu, Jun-Wu Zhang
ABSTRACT
RNA binding proteins (RBPs)-mediated post-transcriptional control has been implicated in influencing various aspects of RNA metabolism and playing important roles in mammalian development and pathological diseases. However, the functions of specific RBPs and the molecular mechanisms through which they act in monocyte/macrophage differentiation remain to be determined. In this study, through bioinformatics analysis and experimental validation, we identify that ZFP36L1, a member of ZFP36 zinc finger protein family, exhibits significant decrease in acute myeloid leukemia (AML) patients compared with normal controls and remarkable time-course increase during monocyte/macrophage differentiation of PMA-induced THP-1 and HL-60 cells as well as induction culture of CD34(+) hematopoietic stem/progenitor cells (HSPCs). Lentivirus-mediated gain and loss of function assays demonstrate that ZFP36L1 acts as a positive regulator to participate in monocyte/macrophage differentiation. Mechanistic investigation further reveals that ZFP36L1 binds to the CDK6 mRNA 3'untranslated region bearing adenine-uridine rich elements and negatively regulates the expression of CDK6 which is subsequently demonstrated to impede the in vitro monocyte/macrophage differentiation of CD34(+) HSPCs. Collectively, our work unravels a ZFP36L1-mediated regulatory circuit through repressing CDK6 expression during monocyte/macrophage differentiation, which may also provide a therapeutic target for AML therapy.
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Anti-ZFP36L1 Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.