Merck
  • Natural and induced antibodies contribute to differential susceptibility to secondary cystic echinococcosis of Balb/c and C57Bl/6 mice.
Natural and induced antibodies contribute to differential susceptibility to secondary cystic echinococcosis of Balb/c and C57Bl/6 mice.
Immunobiology (2015-08-05)
Gustavo Mourglia-Ettlin, Marcela Cucher, Paula Arbildi, Mara Rosenzvit, Sylvia Dematteis
ABSTRACT
Antibodies are key immune players in several helminth infections and animal models have been central for the identification of their mechanisms of protection. Murine secondary cystic echinococcosis is a useful model for studying Echinococcus granulosus immunobiology, being the immune profile mounted by the experimental host a determinant of parasite success or failure in infection establishment. In the present study, we analyzed infection outcome using Balb/c and C57Bl/6 mice strains, and compared their antibody responses in terms of quality and intensity. Our results showed that Balb/c is a highly susceptible strain to secondary cystic echinococcosis, while C57Bl/6 mice are quite resistant. Moreover, significant differences between strains were observed in natural and induced antibodies recognizing E. granulosus antigens, both at the systemic and peritoneal levels. Natural cross-reacting IgM, IgG2b and IgG3 antibodies were detected in sera from both strains but with different intensities, and - remarkably - natural IgG2b showed to be an intrinsic correlate of protection in both mice strains. Interestingly, naïve C57Bl/6 serum displayed a higher protoscolicidal activity, and heterologous - but not homologous - transference of C57Bl/6 naïve serum into Balb/c mice, significantly reduced their infection susceptibility. In the peritoneal cavity, different levels of natural cross-reacting IgM and IgG3 antibodies were detected in both mice strains, while cross-reacting IgG2b was detected only in C57Bl/6 mice. On the other hand, infected mice from both strains developed isotype-mixed antibody responses, with Balb/c mice biasing their response towards high avidity IgG1 and C57Bl/6 mice showing a predominance of mixed IgM/IgG2c/IgG2b/IgG3. In this regard, IgG1 levels showed to be a correlate of susceptibility in both mice strains. In conclusion, our results suggest that antibodies - either natural or induced - play a role in the susceptibility degree to murine secondary cystic echinococcosis.
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, Vetec, reagent grade, 98%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)