Merck
The calpain-suppressing effects of olesoxime in Huntington's disease.
Rare diseases (Austin, Tex.) (2016-05-04)
Jonasz J Weber, Midea M Ortiz Rios, Olaf Riess, Laura E Clemens, Huu P Nguyen
ABSTRACT
Olesoxime, a small molecule drug candidate, has recently attracted attention due to its significant beneficial effects in models of several neurodegenerative disorders including Huntington's disease. Olesoxime's neuroprotective effects have been assumed to be conveyed through a direct, positive influence on mitochondrial function. In a long-term treatment study in BACHD rats, the latest rat model of Huntington's disease, olesoxime revealed a positive influence on mitochondrial function and improved specific behavioral and neuropathological phenotypes. Moreover, a novel target of the compound was discovered, as olesoxime was found to suppress the activation of the calpain proteolytic system, a major contributor to the cleavage of the disease-causing mutant huntingtin protein into toxic fragments, and key player in degenerative processes in general. Results from a second model of Huntington's disease, the Hdh (Q111) knock-in mouse, confirm olesoxime's calpain-suppressing effects and support the therapeutic value of olesoxime for Huntington's disease and other disorders involving calpain overactivation.
Sigma-Aldrich
Anti-Huntingtin Protein Antibody, a.a. 181-810, clone 1HU-4C8, ascites fluid, clone 1HU-4C8, Chemicon®
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-α-Tubulin Mouse mAb (DM1A), liquid, clone DM1A, Calbiochem®
Sigma-Aldrich
Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2, ascites fluid, clone 5TF1-1C2, Chemicon®
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid