Merck
  • BAG5 Interacts with DJ-1 and Inhibits the NeuroprotectiveEffects of DJ-1 to Combat Mitochondrial Oxidative Damage
BAG5 Interacts with DJ-1 and Inhibits the NeuroprotectiveEffects of DJ-1 to Combat Mitochondrial Oxidative Damage
Oxidative Medicine and Cellular Longevity (2017)
Li-xia Qin