Merck
  • NANOG gene expression is regulated by the ETS transcription factor ETV4 in human embryonic carcinoma NCCIT cells.
NANOG gene expression is regulated by the ETS transcription factor ETV4 in human embryonic carcinoma NCCIT cells.
Biochemical and biophysical research communications (2017-04-17)
Sung-Won Park, Hyun-Jin Do, Wonbin Choi, Hyuk Song, Hak-Jae Chung, Jae-Hwan Kim
ABSTRACT
We demonstrated that ETV4 is a transcriptional activator of the NANOG gene in human embryonic carcinoma NCCIT cells. The endogenous expression of NANOG and ETV4 in naïve cells was significantly down-regulated upon differentiation and by shRNA-mediated knockdown of ETV4. NANOG transcription was significantly upregulated by ETV4 overexpression. A putative ETS binding site (EBS) is present in the region (-285 to -138) of the proximal promoter. Site-directed mutagenesis of the putative EBS (
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human ETV4