Merck
  • Overexpression of STIM1 in neurons in mouse brain improves contextual learning and impairs long-term depression.
Overexpression of STIM1 in neurons in mouse brain improves contextual learning and impairs long-term depression.
Biochimica et biophysica acta (2016-12-04)
Łukasz Majewski, Filip Maciąg, Paweł M Boguszewski, Iga Wasilewska, Grzegorz Wiera, Tomasz Wójtowicz, Jerzy Mozrzymas, Jacek Kuznicki
ABSTRACT
STIM1 is an endoplasmic reticulum calcium sensor that is involved in several processes in neurons, including store-operated calcium entry. STIM1 also inhibits voltage-gated calcium channels, such as Ca