Merck
  • Characterization of smokeless powders using multiplexed collision-induced dissociation mass spectrometry and chemometric procedures.
Characterization of smokeless powders using multiplexed collision-induced dissociation mass spectrometry and chemometric procedures.
Forensic science international (2017-01-16)
Kristen L Reese, A Daniel Jones, Ruth Waddell Smith
ABSTRACT
This work demonstrates a non-targeted mass spectrometry approach for identification of organic compounds in smokeless powders. Unburned powders were removed from various commercial ammunitions of different brand, primer composition, caliber, and age. The unburned powders and corresponding fired residues were analyzed by liquid chromatography-atmospheric pressure chemical ionization-time-of-flight mass spectrometry (LC-APCI-TOFMS). Multiplexed collision-induced dissociation was performed at increasing collision potentials resulting in successive fragmentation that provided structural information for compound identification in a non-targeted manner. Nine compounds were identified in the powders, including akardite II, ethyl centralite, diphenylamine, N-nitrosodiphenylamine, and dibutyl phthalate. Multivariate statistical procedures were performed to first investigate association and discrimination of the unburned powders. Principal components analysis (PCA) of the chemical profiles suggested nine distinct groups of powders, according to the dominant organic compounds present. The clusters formed in hierarchical cluster analysis (HCA) were mostly in agreement with PCA groupings although HCA provided a metric to quantify the similarity. Finally, association of the fired residue to the corresponding unburned powder was possible although the success was highly dependent on the composition of the unburned powder and the extent of compound depletion as a result of firing.
Sigma-Aldrich
1,3-Diethyl-1,3-diphenylurea, 99%