Merck
  • A Dioxygenase Catalyzes Steroid 16α-Hydroxylation in Steroidal Glycoalkaloid Biosynthesis.
A Dioxygenase Catalyzes Steroid 16α-Hydroxylation in Steroidal Glycoalkaloid Biosynthesis.
Plant physiology (2017-07-30)
Masaru Nakayasu, Naoyuki Umemoto, Kiyoshi Ohyama, Yoshinori Fujimoto, Hyoung Jae Lee, Bunta Watanabe, Toshiya Muranaka, Kazuki Saito, Yukihiro Sugimoto, Masaharu Mizutani
ABSTRACT
Steroidal glycoalkaloids (SGAs) are toxic specialized metabolites that are found in the Solanaceae. Potato (
Sigma-Aldrich
Solanidine, ≥97.0% (HPLC)
Sigma-Aldrich
22(R)-Hydroxycholesterol, ≥98%
Sigma-Aldrich
22(S)-Hydroxycholesterol