Merck
  • Direct Binding of the Corrector VX-809 to Human CFTR NBD1: Evidence of an Allosteric Coupling between the Binding Site and the NBD1:CL4 Interface.
Direct Binding of the Corrector VX-809 to Human CFTR NBD1: Evidence of an Allosteric Coupling between the Binding Site and the NBD1:CL4 Interface.
Molecular pharmacology (2017-05-27)
Rhea P Hudson, Jennifer E Dawson, P Andrew Chong, Zhengrong Yang, Linda Millen, Philip J Thomas, Christie G Brouillette, Julie D Forman-Kay
ABSTRACT
Understanding the mechanism of action of modulator compounds for the cystic fibrosis transmembrane conductance regulator (CFTR) is key for the optimization of therapeutics as well as obtaining insights into the molecular mechanisms of CFTR function. We demonstrate the direct binding of VX-809 to the first nucleotide-binding domain (NBD1) of human CFTR. Disruption of the interaction between C-terminal helices and the NBD1 core upon VX-809 binding is observed from chemical shift changes in the NMR spectra of residues in the helices and on the surface of
Sigma-Aldrich
BIO-Acetoxime, ≥98% (HPLC)