Merck
  • The use of serine protease from Yarrowia lipolytica yeast in the production of biopeptides from denatured egg white proteins.
The use of serine protease from Yarrowia lipolytica yeast in the production of biopeptides from denatured egg white proteins.
Acta biochimica Polonica (2017-04-08)
Marta Pokora, Aleksandra Zambrowicz, Agnieszka Zabłocka, Anna Dąbrowska, Marek Szołtysik, Konrad Babij, Ewelina Eckert, Tadeusz Trziszka, Józefa Chrzanowska
ABSTRACT
Deriving non-conventional enzymes from cheaper sources than those used for commercially available enzymes may result in the production of hydrolysates with beneficial features, while drastically reducing the cost of hydrolysis. This is especially significant for enzymatic hydrolysis as a method of protein waste utilization. We have previously described the ability of non-commercial serine protease from Yarrowia lipolytica yeast to produce/release bioactive peptides from egg white protein by-products (EP). The enzymatic hydrolysis of EP was carried out for 24 h using the serine protease at an enzyme: substrate ratio of 1:30 (w/w). The obtained hydrolysate was characterized by protein degradation of 38% and also exhibited an antioxidant and cytokine-inducing activity. The isolation procedure (ultrafiltration and RP-HPLC) of bioactive peptides from the EP hydrolysate provided peptide fractions with significant antioxidant and ACE inhibitory activities. Three homogeneous and three heterogeneous peptide fractions were identified using MALDI-TOF/MS and the Mascot Search Results database. The peptides, mainly derived from ovalbumin, were composed of 2-19 amino-acid residues. We have thus demonstrated a novel ability of serine protease from Y. lipolytica to release biopeptides from an EP by-product.
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
Angiotensin Converting Enzyme from rabbit lung, ≥2.0 units/mg protein (modified Warburg-Christian)