Merck
  • Calreticulin regulates TGF-β1-induced epithelial mesenchymal transition through modulating Smad signaling and calcium signaling.
Calreticulin regulates TGF-β1-induced epithelial mesenchymal transition through modulating Smad signaling and calcium signaling.
The international journal of biochemistry & cell biology (2017-08-06)
Yanjiao Wu, Xiaoli Xu, Lunkun Ma, Qian Yi, Weichao Sun, Liling Tang
ABSTRACT
As a Ca
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
MISSION® esiRNA, targeting human CALR3