Merck
  • The Relationship among Tyrosine Decarboxylase and Agmatine Deiminase Pathways in
The Relationship among Tyrosine Decarboxylase and Agmatine Deiminase Pathways in
Frontiers in microbiology (2017-11-23)
Marta Perez, Victor Ladero, Beatriz Del Rio, Begoña Redruello, Anne de Jong, Oscar Kuipers, Jan Kok, M Cruz Martin, Maria Fernandez, Miguel A Alvarez
ABSTRACT
Enterococci are considered mainly responsible for the undesirable accumulation of the biogenic amines tyramine and putrescine in cheeses. The biosynthesis of tyramine and putrescine has been described as a species trait in
Sigma-Aldrich
L-2-Aminoadipic acid, ≥98% (TLC)