Merck
  • Heme oxygenase-1 directly binds STAT3 to control the generation of pathogenic Th17 cells during neutrophilic airway inflammation.
Heme oxygenase-1 directly binds STAT3 to control the generation of pathogenic Th17 cells during neutrophilic airway inflammation.
Allergy (2017-06-06)
X L Lin, J J Lv, J Lv, C X Di, Y J Zhang, T Zhou, J L Liu, Z W Xia
ABSTRACT
Specific JAK/STAT pathways play a critical role in the functional differentiation of distinct Th subsets. Previously, we showed that HO-1, a stress-inducible protein, inhibits Th17 cell differentiation and alleviates neutrophilic airway inflammation, but the responsible molecular basis remains unclear. We employed Th17-skewing differentiation and NEA mouse models to study the role of HO-1 in regulating IL-6-STAT3-RORγt/SOCS3 signaling pathway to control Th17 cell-mediated neutrophilic airway inflammation. The levels of cytokines and expressions of relative signaling molecules were measured by ELISA, western blot, and qPCR, respectively. Frequency of CD4 Here, we show that hemin-induced HO-1 overexpression is required to mediate this process. Specifically, HO-1 decreased STAT3 phosphorylation but not IL-6R/IL-23R expression or JAK1/JAK2 activation in CD4 Our data suggest that HO-1 exerts its inhibitory effect on Th17 cell differentiation by directly associating and blocking STAT3 phosphorylation. We speculate that hemin may be a potential therapeutic candidate for the treatment of other types of immune and pulmonary inflammatory-related diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human HMOX1