Merck
  • Sclerostin and bone metabolism markers in hyperthyroidism before treatment and interrelations between them.
Sclerostin and bone metabolism markers in hyperthyroidism before treatment and interrelations between them.
Acta biochimica Polonica (2017-10-24)
İlker Sarıtekin, Şerefden Açıkgöz, Taner Bayraktaroğlu, Fatih Kuzu, Murat Can, Berrak Güven, Görkem Mungan, Çağatay Büyükuysal, Selda Sarıkaya
ABSTRACT
Sclerostin, which is a glycoprotein produced by osteocytes, reduces the formation of bones by inhibiting the Wnt signal pathway. Thyroid hormones are related with Wnt signal pathway and it has been reported that increased thyroid hormones in hyperthyroidism fasten epiphysis maturation in childhood, and increase the risk of bone fractures by stimulating the bone loss in adults. The aim of this study was to examine the sclerostin serum levels, the relation between sclerostin and thyroid hormones as well as the biochemical markers of the bone metabolism in patients with hyperthyroidism (including multinodular goiter and Graves' disease), whose treatments have not started yet. No difference was found in the serum sclerostin levels between the hyperthyroidism group (n=24) and the control group (n=24) (p=0.452). The serum osteocalcin levels and 24-hour urinary phosphorus excretion were found to be higher in the hyperthyroid group than in the control group (p<0.001, p=0.009). A positive correlation was determined between the sclerostin and bone alkaline phosphatase levels (p<0.001); a negative correlation between the osteocalcin and thyroid stimulating hormone (TSH) (p<0.05); a positive correlation between the osteocalcin and thyroid hormones (FT