Merck
  • Altered rPrP substrate structures and their influence on real-time quaking induced conversion reactions.
Altered rPrP substrate structures and their influence on real-time quaking induced conversion reactions.
Protein expression and purification (2017-10-17)
Keding Cheng, Angela Sloan, Brooks Waitt, Robert Vendramelli, Debra Godal, Sharon L R Simon, Joe O'Neil, Michael Carpenter, Dave Jackson, Jane Eastlake, Gary Mallinson, J David Knox
ABSTRACT
Bacterially-produced recombinant prion protein (rPrP) has traditionally been used for in vitro fibrillation assays and reagent development for prion disease research. In recent years, it has also been used as a substrate for real-time quaking-induced conversion (RT-QuIC), a very sensitive method of detecting the presence of the misfolded, disease-associated isoform of the prion protein (PrP This study examines the effects of two different rPrP refolding protocols on the production, QuIC performance, and structure characteristics of two constructs of rPrP commonly used in QuIC: full length hamster and a sheep-hamster chimeric rPrP. Under the described conditions, the best performing substrate was the chimeric sheep-hamster rPrP produced by shorter guanidine-HCl exposure and faster gradient elution. The observation that different rPrP production protocols influence QuIC performance indicates that caution should be exercised when comparing inter-laboratory QuIC results.
Sigma-Aldrich
N-Oleoylethanolamine, ~98% (TLC)