MilliporeSigma
1.73636
Move DC Nxt Hand-held Colorimeter
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41116116
NACRES:
SE.11
eCl@ss:
40050204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
product line
Spectroquant®
technique(s)
photometry: suitable
application(s)
disinfection control
compatibility
for use with Spectroquant® test kits
storage temp.
15-25°C
General description
Spectroquant®
The Spectroquant® Move DC Nxt mobile colorimeter is suitable for the detection of five essential parameters of water analysis: chlorine, ozone, chlorine dioxide, cyanuric acid and pH. This handy, battery-operated instrument comes with a carrying case and is designed for in-process and field work.
It accelerates disinfection control workflows with rapid analysis, enhanced memory and a user-friendly Dot Matrix UI with icons and animation. The device offers a language-free UI, clear status indicators (green/red backlight), wired data transfer, differentiated measurements and a built-in Favourite list for quick method access.
Move DC Nxt Data Transfer Software (Photometer Data Transfer Tool)
Legal Information
SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Visit the Document Library