Skip to Content
Merck

216577

Amberlite IRC-50S

400 μm (wet)

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

CAS Number:
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Amberlite IRC-50S, 400 μm (wet)

vapor density

<1 (vs air)

vapor pressure

17 mmHg ( 20 °C)

autoignition temp.

800 °F

particle size

400 μm (wet)

Features and Benefits

Weakly acidic resin with carboxylic acid functionality which is useful in catalytic applications.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service