Merck
  • Evaluating cell-surface expression and measuring activation of mammalian odorant receptors in heterologous cells.
Evaluating cell-surface expression and measuring activation of mammalian odorant receptors in heterologous cells.
Nature protocols (2008-09-06)
Hanyi Zhuang, Hiroaki Matsunami
ABSTRACT
A fundamental question in olfaction is which odorant receptors (ORs) are activated by a given odorant. A major roadblock to investigating odorant-OR relationships in mammals has been the inability to express ORs in heterologous cells suitable for screening active ligands for ORs. The discovery of the receptor-transporting protein family has facilitated the effective cell-surface expression of ORs in heterologous cells. The establishment of a robust heterologous expression system for mammalian ORs facilitates the high-throughput 'deorphanization' of these receptors by matching them to their cognate ligands. This protocol details the method used for evaluating the cell-surface expression and measuring the functional activation of ORs of transiently expressed mammalian ORs in HEK293T cells. The stages of OR cell-surface expression include cell culture preparation, transfer of cells, transfection, immunocytochemistry or flow cytometry, odorant stimulation and luciferase assay. This protocol can be completed in a period of 3 d from the transfer of cells to cell-surface expression detection and/or measurement of functional activation.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)