Merck
  • A dietary fiber-deprived gut microbiota degrades the colonic mucus barrier and enhances pathogen susceptibility
A dietary fiber-deprived gut microbiota degrades the colonic mucus barrier and enhances pathogen susceptibility
Cell (2016)
Desai MS, et al.