Merck
  • Attenuation of hedgehog/GLI signaling by NT1721 extends survival in pancreatic cancer.
Attenuation of hedgehog/GLI signaling by NT1721 extends survival in pancreatic cancer.
Journal of experimental & clinical cancer research : CR (2019-10-30)
Claudia M Kowolik, Min Lin, Jun Xie, Larry E Overman, David A Horne
ABSTRACT
Pancreatic cancer is one of the most lethal malignancies due to frequent late diagnosis, aggressive tumor growth and metastasis formation. Continuously raising incidence rates of pancreatic cancer and a lack of significant improvement in survival rates over the past 30 years highlight the need for new therapeutic agents. Thus, new therapeutic agents and strategies are urgently needed to improve the outcome for patients with pancreatic cancer. Here, we evaluated the anti-tumor activity of a new natural product-based epidithiodiketopiperazine, NT1721, against pancreatic cancer. We characterized the anticancer efficacy of NT1721 in multiple pancreatic cancer cell lines in vitro and in two orthotopic models. We also compared the effects of NT1721 to clinically used hedgehog inhibitors and the standard-of-care drug, gemcitabine. The effect of NT1721 on hedgehog/GLI signaling was assessed by determining the expression of GLI and GLI target genes both in vitro and in vivo. NT1721 displayed IC50 values in the submicromolar range in multiple pancreatic cancer cell lines, while largely sparing normal pancreatic epithelial cells. NT1721 attenuated hedgehog/GLI signaling through downregulation of GLI1/2 transcription factors and their downstream target genes, which reduced cell proliferation and invasion in vitro and significantly decreased tumor growth and liver metastasis in two preclinical orthotopic mouse models of pancreatic cancer. Importantly, treatment with NT1721 significantly improved survival times of mice with pancreatic cancer compared to the standard-of-care drug, gemcitabine. Favorable therapeutics properties, i.e. 10-fold lower IC50 values than clinically used hedgehog inhibitors (vismodegib, erismodegib), a 90% reduction in liver metastasis and significantly better survival times compared to the standard-of-care drug, gemcitabine, provide a rational for testing NT1721 in the clinic either as a single agent or possibly in combination with gemcitabine or other therapeutic agents in PDAC patients overexpressing GLI1/2. This could potentially result in promising new treatment options for patients suffering from this devastating disease.
Sigma-Aldrich
Anti-Protein patched homolog 1 Antibody, from rabbit, purified by affinity chromatography
Pricing and availability is not currently available.