Merck
  • Genipin inhibits rotavirus-induced diarrhea by suppressing viral replication and regulating inflammatory responses.
Genipin inhibits rotavirus-induced diarrhea by suppressing viral replication and regulating inflammatory responses.
Scientific reports (2020-09-29)
Jong-Hwa Kim, Kiyoung Kim, Wonyong Kim
ABSTRACT
Rotavirus is the leading cause of acute gastroenteritis among young children worldwide. However, agents specifically designed to treat rotavirus infection have not been developed yet. In this study, the anti-rotavirus and anti-inflammatory effects of genipin, a chemical compound found in the fruit of Gardenia jasminoides, were evaluated. Genipin had an antiviral effect against the human rotavirus Wa and SA-11 strains in vitro, and it inhibited two distinct stages of the viral replication cycle: attachment and penetration (early stage) in pre-treatment and assembly and release (late stage) in post-treatment. Additionally, genipin downregulated nitric oxide synthase and pro-inflammatory cytokines in lipopolysaccharide-stimulated RAW264.7 cells and rotavirus-infected Caco-2 cells. Oral administration of genipin before and after viral infection with the murine rotavirus epidemic diarrhea of infant mice strain led to a reduced duration of diarrhea and faecal viral shedding and to decreased destruction of the enteric epithelium. Genipin could have potential as a natural compound with preventive and therapeutic effects against infection and colitis caused by rotavirus.
Sigma-Aldrich
Genipin, ≥98% (HPLC), powder
Pricing and availability is not currently available.