Merck
  • Benzoic acid and pyridine derivatives as inhibitors of Trypanosoma cruzi trans-sialidase.
Benzoic acid and pyridine derivatives as inhibitors of Trypanosoma cruzi trans-sialidase.
Bioorganic & medicinal chemistry (2007-01-16)
João Neres, Pascal Bonnet, Philip N Edwards, Pravin L Kotian, Alejandro Buschiazzo, Pedro M Alzari, Richard A Bryce, Kenneth T Douglas
ABSTRACT
Benzoic acid and pyridine derivatives inhibit recombinant trans-sialidase from Trypanosoma cruzi with I50 values between 0.4 and 1mM. The best compounds, 4-acetylamino-3-hydroxymethylbenzoic acid and 5-acetylamino-6-aminopyridine-2-carboxylic acid, provide new leads to inhibitors not containing the synthetically complex sialic acid structure. The weak inhibition by such compounds contrasts with their much stronger inhibition of neuraminidase from Influenza virus.
Sigma-Aldrich
4-Acetamidobenzoic acid, 98%
Pricing and availability is not currently available.