Merck
  • Cohesin promotes HSV-1 lytic transcription by facilitating the binding of RNA Pol II on viral genes.
Cohesin promotes HSV-1 lytic transcription by facilitating the binding of RNA Pol II on viral genes.
Virology journal (2021-01-25)
Xin Li, Yafen Yu, Fengchao Lang, Guijun Chen, Erlin Wang, Lihong Li, Zhuoran Li, Liping Yang, Xia Cao, Nigel W Fraser, Jumin Zhou
ABSTRACT
Herpes Simplex Virus type I (HSV-1) is a large double-stranded DNA virus that enters productive infection in epithelial cells and reorganizes the host nucleus. Cohesin, a major constituent of interphase and mitotic chromosomes comprised of SMC1, SMC3, and SCC1 (Mcd1/Rad21), SCC3 (SA1/SA2), have diverse functions, including sister chromatid cohesion, DNA double-stranded breaks repair, and transcriptional control. Little is known about the role of cohesin in HSV-1 lytic infection. We measured the effect on HSV-1 transcription, genome copy number, and viral titer by depleting cohesin components SMC1 or Rad21 using RNAi, followed by immunofluorescence, qPCR, and ChIP experiments to gain insight into cohesin's function in HSV-1 transcription and replication. Here, we report that cohesion subunits SMC1 and Rad21 are recruited to the lytic HSV-1 replication compartment. The knockdown results in decreased viral transcription, protein expression, and maturation of viral replication compartments. SMC1 and Rad21 knockdown leads to the reduced overall RNA pol II occupancy level but increased RNA pol II ser5 phosphorylation binding on viral genes. Consistent with this, the knockdown increased H3K27me3 modification on these genes. These results suggest that cohesin facilitates HSV-1 lytic transcription by promoting RNA Pol II transcription activity and preventing chromatin's silencing on the viral genome.
Sigma-Aldrich
ChIPAb+ CTCF - ChIP Validated Antibody and Primer Set, clone 1.2.1.5.4, from mouse
Pricing and availability is not currently available.