Merck
  • TRIB3 Mediates Fibroblast Activation and Fibrosis though Interaction with ATF4 in IPF.
TRIB3 Mediates Fibroblast Activation and Fibrosis though Interaction with ATF4 in IPF.
International journal of molecular sciences (2022-12-24)
Lan Wang, Wenyu Zhao, Cong Xia, Zhongzheng Li, Weiming Zhao, Kai Xu, Ningdan Wang, Hui Lian, Ivan O Rosas, Guoying Yu
ABSTRACT
Idiopathic pulmonary fibrosis (IPF) is a fatal interstitial lung disease characterized by fibroblast activation, excessive deposition of extracellular matrix, and progressive scarring; the pathogenesis remains elusive. The present study explored the role of Tribbles pseudokinase 3 (TRIB3), a well-known stress and metabolic sensor, in IPF. TRIB3 is down-regulated in the lungs of IPF patients in comparison to control subjects. Deficiency of TRIB3 markedly inhibited A549 epithelial cells' proliferation and migration, significantly reducing wound healing. Conversely, overexpression of TRIB3 promoted A549 cell proliferation and transmigration while it inhibited its apoptosis. Meanwhile, overexpressed TRIB3 inhibited fibroblast activation and decreased ECM synthesis and deposition in MRC5 cells. TRIB3 attenuated pulmonary fibrosis by negative regulation of ATF4, while TRIB3 expression markedly inhibited ATF4 promoter-driven transcription activity and down-regulated ATF4 expression. A co-culture system showed that TRIB3 is important to maintain the normal epithelial-mesenchymal crosstalk and regulate fibroblast activation. Taken together, our data suggested that an axis of TRIB3-ATF4 is a key mediator in IPF which might be a potential target for fibroproliferative lung disease treatment.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)